Troemner Class-6 Avoirdupois Cast Iron Slotted Weights
Use in a variety of applications such as pressure, torque, and tensile strength testing
  • Use several slotted weights together to build up from a minimum weight to a maximum test load
  • A durable coat of paint protects each weight from rusting and chipping
  • Color coded – silver for avoirdupois
Cast Iron Slotted Weights are used in a variety of applications such as pressure, torque, and tensile strength testing. Cast Iron Slotted Weights are typically used with a hanger that also has its weight calibrated so the hanger can be used as part of the overall weight under test. Several Cast Iron Slotted Weights may be used together to build up from a minimum weight to a maximum test load. The hanger weight selected should be able to accommodate the total load needed.
Cast Iron Slotted Weights above 2 kg/5 lb have adjusting cavities. Weights below 2 kg/5 lb are adjusted by removing material from the underside of the weight. Each weight has its nominal value cast into the topside of the weight.
9,900.00 - ₹66,900.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (lbs) Description
Availability Pricing
Troemner 9498 50 lb, 10-1/4" Dia, Avoirdupois Slotted Weights WW-11966-59 Mfr # 9498
COMPARE
None 50 50 lb, 10-1/4\" Dia, Avoirdupois Slotted Weights
₹50,700.00INR / Each
Troemner 9497 25 lb, 10-1/4" Dia, Avoirdupois Slotted Weights WW-11966-60 Mfr # 9497
COMPARE
None 25 25 lb, 10-1/4\" Dia, Avoirdupois Slotted Weights
₹36,900.00INR / Each
Troemner 9496 20 lb, 8" Dia, Avoirdupois Slotted Weights WW-11966-61 Mfr # 9496
COMPARE
None 20 20lb, 8\" Dia, Avoirdupois Slotted Weights
₹24,400.00INR / Each
Troemner 9494 10 lb, 6" Dia, Avoirdupois Slotted Weights WW-11966-62 Mfr # 9494
COMPARE
None 10 10lb, 6\" Dia, Avoirdupois Slotted Weights
₹19,600.00INR / Each
Troemner 9492 5 lb, 5" Dia, Avoirdupois Slotted Weights WW-11966-63 Mfr # 9492
COMPARE
None 5 5lb, 5\" Dia, Avoirdupois Slotted Weights
₹15,900.00INR / Each
Troemner 9490 2 lb, 5" Dia, Avoirdupois Slotted Weights WW-11966-64 Mfr # 9490
COMPARE
None 2 2lb, 5\" Dia, Avoirdupois Slotted Weights
₹13,000.00INR / Each
Troemner 9488 1 lb, 5" Dia, Avoirdupois Slotted Weights WW-11966-65 Mfr # 9488
COMPARE
None 1 1lb, 5\" Dia, Avoirdupois Slotted Weights
₹12,600.00INR / Each
Troemner 9486 0.5 lb, 3-3/4" Dia, Avoirdupois Slotted Weights WW-11966-67 Mfr # 9486
COMPARE
None 0.5 0.5lb, 3-3/4\" Dia, Avoirdupois Slotted Weights
₹9,900.00INR / Each
Troemner 9498T 50lb 10-1/4" Dia, Avoirdupois Slotted Weights, with Traceable Certificate, WW-11966-70 Mfr # 9498T
COMPARE
NIST 50 50lb, 10-1/4\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate,
₹66,900.00INR / Each
Troemner 9497T 25Lb 10-1/4" Dia, Avoirdupois Slotted Weights, with Traceable Certificate, WW-11966-71 Mfr # 9497T
COMPARE
NIST 25 25lb, 10-1/4\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate,
₹50,700.00INR / Each
Troemner 9496T 20lb 8" Dia, Avoirdupois Slotted Weights, with Traceable Certificate, WW-11966-72 Mfr # 9496T
COMPARE
NIST 20 20lb, 8\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate,
₹37,700.00INR / Each
Troemner 9494T 10lb 6" Dia, Avoirdupois Slotted Weights, with Traceable Certificate WW-11966-73 Mfr # 9494T
COMPARE
NIST 10 10lb, 6\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate
₹32,800.00INR / Each
Troemner 9492T 5Lb 5" Dia, Avoirdupois Slotted Weights, with Traceable Certificate WW-11966-74 Mfr # 9492T
COMPARE
NIST 5 5lb, 5\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate
₹27,500.00INR / Each
Troemner 9490T 2Lb 5" Dia, Avoirdupois Slotted Weights, with Traceable Certificate WW-11966-75 Mfr # 9490T
COMPARE
NIST 2 2lb, 5\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate
₹23,200.00INR / Each
Troemner 9488T 1Lb 5" Dia, Avoirdupois Slotted Weights, with Traceable Certificate WW-11966-76 Mfr # 9488T
COMPARE
NIST 1 1lb, 5\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate
₹21,800.00INR / Each
Troemner 9486T 0.5Lb 3-3/4" Dia, Avoirdupois Slotted Weights, with Traceable Certificate WW-11966-77 Mfr # 9486T
COMPARE
NIST 0.5 0.5lb, 3-3/4\" Dia, Avoirdupois Slotted Weights, with Traceable Certificate
₹20,800.00INR / Each
Showing 1- 16 of 16