TriForest Sterile Square Media Bottles, PETG
Gamma sterilized, shatterproof bottles with tamper-evident caps
  • PETG is a safe, shatterproof glass alternative
  • Leakproof seals (included) secure samples
  • Complete reference fills—molded-in graduations make it easy
  • Gamma irradiated for SAL 10–6 sterility
Store solutions and critical fluids in these sturdy, sterile bottles. The square shape saves shelf space—place bottles closer together.
Bottles come in case quantities and contain smaller packs within. Choose from bagged bottles or bottles packaged in trays. Each case is specially packaged to maintain sterility with multibarrier packaging.
57,600.00 - ₹87,900.00INR / Case of 288
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Special Features
Availability Pricing
TriForest BGC0030S Square Media Bottle, Sterile, 30 mL, PETG; 24/PK, 288/CS 0654200 Mfr # BGC0030S-A
COMPARE
30 Packed in bags
₹80,100.00INR / Case of 288
TriForest BGC0060S Square Media Bottle, Sterile, 60 mL, PETG; 24/PK, 192/CS 0654206 Mfr # BGC0060S-A
COMPARE
60 Packed in bags
₹87,900.00INR / Case of 192
TriForest BGC0125S Square Media Bottle, Sterile, 125 mL, PETG; 24/PK, 144/CS 0654207 Mfr # BGC0125S-A
COMPARE
125 Packed in bags
₹86,500.00INR / Case of 144
TriForest BGC0250S Square Media Bottle, Sterile, 250 mL, PETG; 24/PK, 96/CS 0654208 Mfr # BGC0250S-A
COMPARE
250 Packed in bags
₹75,900.00INR / Case of 96
TriForest BGC0500S Square Media Bottle, Sterile, 500 mL, PETG; 12/PK, 72/CS 0654209 Mfr # BGC0500S-A
COMPARE
500 Packed in bags
₹72,000.00INR / Case of 72
TriForest BGC1000S Square Media Bottle, Sterile, 1000 mL, PETG; 12/PK, 48/CS 0654210 Mfr # BGC1000S-A
COMPARE
1000 Packed in bags
₹57,600.00INR / Case of 48
TriForest BGC2000S Square Media Bottle, Sterile, 2000 mL, PETG; 6/PK, 24/CS 0654211 Mfr # BGC2000S-A
COMPARE
2000 Packed in bags
₹76,300.00INR / Case of 24
Showing 1- 7 of 7