Trajan Syringes for Rheodyne and Valco
Designed specifically for Rheodyne and Valco injection valves.
  • Metal back flange with flat sides prevents syring rolling
  • White backing strip with black scale markings for accurate reading
  • Metal cap protects syringe barel.
Uses a 22 gauge 5.1cm long needle with a LC needle tops style, no electrotaper.
4,900.00 - ₹29,000.00INR / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 001301 Microvolume Syringe, Fixed LC Needle, 5 uL, 51 mm L, 22 G; 1/Ea 1599905 Mfr # 001301
COMPARE
₹6,700.00INR / Each
Trajan SGE 002301 HPLC Valve Syringe, Fixed LC Needle, 10 uL, 51 mm L, 22 G, LC Tip Needle; 1/Ea 1599947 Mfr # 002301
COMPARE
₹4,900.00INR / Each
Trajan SGE 002313 Removable Needle Syringe with Removable Plunger, 10 uL, 51 mm L, 22 G, LC Needle, 1/Ea 1599948 Mfr # 002313
COMPARE
₹8,000.00INR / Each
Trajan SGE 002315 HPLC Valve Syringe with Fixed LC Needle, 10 uL, 51 mm L, 22 G; 6/Pk 1599949 Mfr # 002315
COMPARE
₹29,000.00INR / Pkg of 6
Trajan SGE 002335 Removable Needle Syringe with Removable Plunger; 10 uL, 51 mm L, 22 G, LC Needle, 1/Ea 1599951 Mfr # 002335
COMPARE
₹6,900.00INR / Each
Trajan SGE 003312 Gas Tight Syringe, Removable LC/Blunt-Tip Needle, 25 uL, 51 mm L; 1/Ea 1600036 Mfr # 003312
COMPARE
₹8,500.00INR / Each
Trajan SGE 004300 Fixed Needle Gas Tight Syringe, 50 uL, 51 mm L, 22 G, LC/Blunt Tip; 1/Ea 1600058 Mfr # 004300
COMPARE
₹5,700.00INR / Each
Trajan SGE 004312 Gas Tight Syringe, Removable LC/Blunt-Tip Needle, 50 uL, 51 mm L, 22 G; 1/Ea 1600059 Mfr # 004312
COMPARE
₹8,000.00INR / Each
Trajan SGE 005300 Fixed Needle LC/Blunt-Tip Needle Syringe, 100 uL, 51 mm L, 22 G; 1/Ea 1600085 Mfr # 005300
COMPARE
₹5,900.00INR / Each
Trajan SGE 005312 Microvloume Syringe, Removable LC/Blunt-Tip Needle, 100 uL, 22 G, 51 mm L, LC tip; 1/Ea 1600086 Mfr # 005312
COMPARE
₹9,000.00INR / Each
Trajan SGE 006300 Fixed Needle Syringe, 250 uL, 51 mm L, 22 G; LC/Blunt Tip Needle; 1/Ea 1600117 Mfr # 006300
COMPARE
₹7,900.00INR / Each
Trajan SGE 006312 Gas Tight Syringe, Removable LC/Blunt-Tip Needle, 250 uL, 51 mm L, 22 G; 1/Ea 1600119 Mfr # 006312
COMPARE
₹9,100.00INR / Each
Trajan SGE 007300 Fixed LC/Blunt-Tip Needle Syringe, 500 uL, 51 mm L, 22 G; 1/Ea 1600142 Mfr # 007300
COMPARE
₹9,600.00INR / Each
Trajan SGE 007310 Microvloume Syringe with Removable Needle, 500 uL, 22 G, 51 mm L, LC/Blunt Tip; 1/Ea 1600143 Mfr # 007310
COMPARE
₹12,600.00INR / Each
Trajan SGE 007312 Removable Needle Gas Tight Syringe, 500 uL, 51 mm L, 22 G, LC/Blunt Tip; 1/Ea 1600144 Mfr # 007312
COMPARE
₹9,200.00INR / Each
Trajan SGE 008105 Gas Tight Syringe, Removable LC/Blunt-Tip Needle, 1 mL, 51 mm L, 22 G; 1/Ea 1600158 Mfr # 008105
COMPARE
₹11,400.00INR / Each
Trajan SGE 008505 Gas Tight Syringe, Removable LC/Blunt-Tip Needle, 2.5 mL, 51 mm L, 22 G; 1/Ea 1600173 Mfr # 008505
COMPARE
₹12,500.00INR / Each
Trajan SGE 003300 Fixed Needle Syringe, 25 uL, 51 mm L, 22 G; LC/Blunt Tip Needle; 1/Ea 1601137 Mfr # 003300
COMPARE
₹6,200.00INR / Each
Showing 1- 18 of 18