Trajan SGE 002313 Removable Needle Syringe with Removable Plunger, 10 uL, 51 mm L, 22 G, LC Needle, 1/Ea
Representative image only.
Trajan SGE 002313 Removable Needle Syringe with Removable Plunger, 10 uL, 51 mm L, 22 G, LC Needle, 1/Ea
Trajan | Mfr: 002313| Item: 1599948
Designed specifically for Rheodyne and Valco injection valves.
₹8,000.00INR / Each
₹8,000.00INR / Each
Product Description
Key Features
  • Metal back flange with flat sides prevents syring rolling
  • White backing strip with black scale markings for accurate reading
  • Metal cap protects syringe barel.
  • 6 month warranty
More About this Item
Uses a 22 gauge 5.1cm long needle with a LC needle tops style, no electrotaper.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Removable Needle Syringe with Removable Plunger, 10 uL, 51 mm L, 22 G, LC Needle, 1/Ea