Trajan Syringe Valves
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
5,600.00 - ₹9,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 031915 Syringes Valve with Luer Lock and Luer Tip, for 5 mL-2 L Syringes; 1/Pk 1600282 Mfr # 031915
COMPARE
₹7,100.00INR / Each
Trajan SGE 038810 Valve Needle, 50 mm L, 23 G, 0.63 mm OD, Cone Tip; 1/Pk 1600462 Mfr # 038810
COMPARE
₹5,600.00INR / Pkg of 1
Trajan SGE 038815 Valve Needles, 50 mm L, 23 G, 0.63 mm OD, Side Hole/Dome Tip; 1/Pk 1600463 Mfr # 038815
COMPARE
₹9,600.00INR / Pkg of 1
Trajan SGE 031910 Push Button with Luer Lock, Compatible with 5 mL-2 L Syringes; 1/Pk 1601157 Mfr # 031910
COMPARE
₹7,100.00INR / Each