Trajan SGE 038810 Valve Needle, 50 mm L, 23 G, 0.63 mm OD, Cone Tip; 1/Pk
Representative image only.
Trajan SGE 038810 Valve Needle, 50 mm L, 23 G, 0.63 mm OD, Cone Tip; 1/Pk
Trajan | Mfr: 038810| Item: 1600462
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹5,600.00INR / Pkg of 1
₹5,600.00INR / Pkg of 1
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • DescriptionValve Needle, 50 mm L, 23 G, 0.63 mm OD, Cone Tip; 1/Pk