Trajan SGE PTV/LVI Inlet Liners
Spot the Difference in the SGE Lineup!
  • Sintered glass beads or powder increases surface area and traps nonvolatile residue
  • Uses baffles or quartz wool to aid in sample vaporization and retain droplets during injection
  • Side hole needles recommended to ensure distribution of sample within the liner
PTV/LVI liners available for Agilent/Gerstel, PerkinElmer, Shimadzu 2010 injector, Thermo Scientific, and Varian/Bruker 1078/1079 injectors
32,300.00 - ₹47,700.00INR / Pkg of 5
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 092210 AC Control Injector Liner, PTV Untreated, Compatible with Agilent Systems, 73 mm L; 5/Pk 1601067 Mfr # 092210
COMPARE
₹47,700.00INR / Pkg of 5
Trajan SGE 092243 LVI Liner, Sintered Glass, Compatible with Agilent Systems, 1.8 mm ID, 71 mm L; 5/Pk 1601085 Mfr # 092243
COMPARE
₹32,300.00INR / Pkg of 5