Trajan SGE 092210 AC Control Injector Liner, PTV Untreated, Compatible with Agilent Systems, 73 mm L; 5/Pk
Representative image only.
Trajan SGE 092210 AC Control Injector Liner, PTV Untreated, Compatible with Agilent Systems, 73 mm L; 5/Pk
Trajan | Mfr: 092210| Item: 1601067
Spot the Difference in the SGE Lineup!
₹47,700.00INR / Pkg of 5
₹47,700.00INR / Pkg of 5
Product Description
Key Features
  • Sintered glass beads or powder increases surface area and traps nonvolatile residue
  • Uses baffles or quartz wool to aid in sample vaporization and retain droplets during injection
  • Side hole needles recommended to ensure distribution of sample within the liner
  • 6 month warranty
More About this Item
PTV/LVI liners available for Agilent/Gerstel, PerkinElmer, Shimadzu 2010 injector, Thermo Scientific, and Varian/Bruker 1078/1079 injectors
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE AC Control Injector Liner, PTV Untreated, Compatible with Agilent Systems, 73 mm L; 5/Pk
  • Warranty6 month warranty