Trajan SGE eVol® Electronic Syringe
Digitally controlled electronic
drive and an analytical
syringe-all in one!
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow-up to 98 steps
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Use for both GC and LC injections
Create a more efficient workflow using this programmable digital syringe. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination.
Calibration and password protection features ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
eVoL XR Electronic Syringe Starter Kit includes:
Handle, universal charger, stand, and three syringes (5 μL, 100 μL, and 1 mL syringes with 5 cm needles) with beveltipped stainless steel needles.
eVoL Electronic Syringe Starter Kit, NMR Edition includes:
Handle, universal charger, stand, and three syringes (5 μL syringe with 11.5 cm needle, 50 μL syringe with 18 cm needle, and 500 μL syringe with 11.5 cm and 18 cm needles) with bevel-tipped stainless steel needles.
14,600.00 - ₹43,900.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Trajan SGE Electronic Syringe: 5µl eVol® Syringe WW-15974-04 Mfr # 2910020
COMPARE
₹15,500.00INR / Each
Trajan SGE Electronic Syringe: 5µl eVol® Syringe supplied without needle WW-15974-05 Mfr # 2910021
COMPARE
₹14,600.00INR / Each
Trajan SGE Electronic Syringe: 50µl eVol® Syringe WW-15974-06 Mfr # 2910022
COMPARE
₹15,500.00INR / Each
Trajan SGE Electronic Syringe: 50µl eVol® Syringe supplied without needle WW-15974-07 Mfr # 2910023
COMPARE
₹14,600.00INR / Each
Trajan SGE Electronic Syringe: 500µl eVol® Syringe WW-15974-08 Mfr # 2910024
COMPARE
₹16,300.00INR / Each
Trajan SGE: EVOL Syringe, 500ul (No Needle) WW-15974-09 Mfr # 2910025
COMPARE
₹15,500.00INR / Each
Trajan SGE: 500UL EVOL MEPS SYRINGE WW-15974-10 Mfr # 2910026
COMPARE
₹19,200.00INR / Each
Trajan SGE: 50UL EVOL MEPS SYRINGE WW-15974-11 Mfr # 2910027
COMPARE
₹18,100.00INR / Each
Trajan SGE Electronic Syringe: 100µl eVol® Syringe for MEPS (new 2011 Firmware required) WW-15974-12 Mfr # 2910028
COMPARE
₹19,200.00INR / Each
Trajan SGE : 100UL EVOL SYRINGE WW-15974-13 Mfr # 2910029
COMPARE
₹16,300.00INR / Each
Trajan SGE Electronic Syringe: 1mL eVol® Syringe WW-15974-14 Mfr # 2910035
COMPARE
₹16,300.00INR / Each
Trajan SGE: 100UL EVOL SYRINGE ( NO NEEDLE) WW-15974-15 Mfr # 2910037
COMPARE
₹15,500.00INR / Each
Trajan SGE Electronic Syringe: 5µl eVol® Syringe pack of 3 WW-15974-16 Mfr # 2910320
COMPARE
₹41,600.00INR / Pkg of 3
Trajan SGE: 50UL EVOL SYRINGE WW-15974-17 Mfr # 2910322
COMPARE
₹41,600.00INR / Pkg of 3
Trajan SGE: 500UL EVOL SYRINGE WW-15974-18 Mfr # 2910324
COMPARE
₹43,900.00INR / Pkg of 3
Trajan SGE Electronic Syringe: 100µl eVol® Syringe pack of 3 (new 2011 Firmware required) WW-15974-19 Mfr # 2910329
COMPARE
₹43,900.00INR / Pkg of 3
Trajan SGE : 1ML EVOL SYRINGE WW-15974-20 Mfr # 2910335
COMPARE
₹43,900.00INR / Pkg of 3
Showing 1- 17 of 17