Trajan SGE: 500UL EVOL MEPS SYRINGE
Representative image only.
Trajan SGE: 500UL EVOL MEPS SYRINGE
Trajan | Mfr: 2910026| Item: 1597410
Digitally controlled electronic
drive and an analytical
syringe-all in one!
₹23,700.00INR / Each
₹23,700.00INR / Each
Product Description
Key Features
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow-up to 98 steps
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Use for both GC and LC injections
More About this Item
Create a more efficient workflow using this programmable digital syringe. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination.
Calibration and password protection features ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
eVoL XR Electronic Syringe Starter Kit includes:
Handle, universal charger, stand, and three syringes (5 μL, 100 μL, and 1 mL syringes with 5 cm needles) with beveltipped stainless steel needles.
eVoL Electronic Syringe Starter Kit, NMR Edition includes:
Handle, universal charger, stand, and three syringes (5 μL syringe with 11.5 cm needle, 50 μL syringe with 18 cm needle, and 500 μL syringe with 11.5 cm and 18 cm needles) with bevel-tipped stainless steel needles.