Trajan Replacement O-Rings
High-performance sealing solutions for GC systems
  • Graphite and Viton materials ensure chemical resistance and high-temperature durability
  • Compatible with leading brands: Agilent, PerkinElmer, and Shimadzu GC systems
  • Designed for tight, leak-free seals in inlet liners and O-ring applications
  • Available in packs of 10 for convenient stocking and replacements
This assortment of sealing rings offers reliable compatibility with popular GC instrument brands including Agilent, PerkinElmer, and Shimadzu. Whether you need inlet liner sealing rings or O-rings, these components are engineered for high-temperature stability and resistance to chemical degradation. With precise outer diameter specifications, they ensure a secure fit and minimal downtime in analytical workflows.
3,400.00 - ₹14,200.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Trajan Graphite Sealing Ring, Compatible with Thermo Scintific Liners, 8 mm ID; 10/Pk 1600711 Mfr # 0726004
COMPARE
Trajan SGE Graphite Sealing Ring, Compatible with Thermo Scintific Liners, 8 mm ID; 10/Pk
₹14,200.00INR / Pkg of 10
Trajan Graphite Inlet Liner Sealing Ring, Compatible with Agilent Systems; 10/Pk 1600712 Mfr # 0726005
COMPARE
Trajan SGE Graphite Inlet Liner Sealing Ring, Compatible with Agilent Systems; 10/Pk
₹6,700.00INR / Pkg of 10
Trajan Graphite Sealing Ring, Compatible with PerkinElmer PSS Liners; 10/Pk 1600752 Mfr # 0726522
COMPARE
Trajan SGE Graphite Sealing Ring, Compatible with Perkin Elmer PSS Liners; 10/Pk
₹3,400.00INR / Pkg of 10
Trajan Viton Inlet Liner Sealing Ring, Compatible with Agilent Liners; 10/Pk 1600754 Mfr # 0726532
COMPARE
Trajan SGE Viton Inlet Liner Sealing Ring, Compatible with Agilent Liners; 10/Pk
₹5,100.00INR / Pkg of 10
Trajan Viton O-Ring, Compatible with Shimadzu, Compatible with 2010 and 2014 Liners, 5 mm OD, 10/Pk 1600755 Mfr # 0726533
COMPARE
Trajan SGE Viton O-Ring, Compatible with Shimadzu, Compatible with 2010 and 2014 Liners, 5 mm OD, 10/Pk
₹5,900.00INR / Pkg of 10
Trajan Viton O' Ring Seal, Compatible with PerkinElmer 6.2 mm OD Liners; 10/Pk 1600756 Mfr # 0726536
COMPARE
Trajan SGE Viton O' Ring Seal, Compatible with Perkin Elmer 6.2 mm OD Liners; 10/Pk
₹5,900.00INR / Pkg of 10