Trajan Viton Inlet Liner Sealing Ring, Compatible with Agilent Liners; 10/Pk
Representative image only.
Trajan Viton Inlet Liner Sealing Ring, Compatible with Agilent Liners; 10/Pk
Trajan | Mfr: 0726532| Item: 1600754
High-performance sealing solutions for GC systems
₹5,100.00INR / Pkg of 10
₹5,100.00INR / Pkg of 10
Product Description
Key Features
  • Graphite and Viton materials ensure chemical resistance and high-temperature durability
  • Compatible with leading brands: Agilent, PerkinElmer, and Shimadzu GC systems
  • Designed for tight, leak-free seals in inlet liners and O-ring applications
  • Available in packs of 10 for convenient stocking and replacements
  • 6 month warranty
More About this Item
This assortment of sealing rings offers reliable compatibility with popular GC instrument brands including Agilent, PerkinElmer, and Shimadzu. Whether you need inlet liner sealing rings or O-rings, these components are engineered for high-temperature stability and resistance to chemical degradation. With precise outer diameter specifications, they ensure a secure fit and minimal downtime in analytical workflows.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Viton Inlet Liner Sealing Ring, Compatible with Agilent Liners; 10/Pk
  • Warranty6 month warranty