Trajan Removeable Cone-Tip Needle Autosampler NanoVolume Syringe Replacement Plungers
Inject down to 50nL with high precision and accuracy.
  • Plunger extends into the needle tip
  • Sample contained only in the needle, no glass contact
Ideal for dispensing very small liquid volumes. When the plunger is depressed, the sample is completely dispensed by the NanoVolume plunger that extends to the tip of the needle.
7,400.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 033057 Replacement Kit, for 0.5 uL Plunger-in-Needle Syringe, 70 mm L, 0.63 mm OD, CT; 1/Ea 1600300 Mfr # 033057
COMPARE
₹7,400.00INR / Each
Trajan SGE 033060 Replacement Kit, for 0.5 uL Plunger-in-Needle Syringe, 70 mm L, 0.63 mm OD, BT; 1/Ea 1600301 Mfr # 033060
COMPARE
₹7,400.00INR / Each
Trajan SGE 034055 Replacement Kit, Compatible with 1 uL Plunger-in-Needle Syringe, 50 mm L, 0.63 mm OD, CT; 1/Ea 1600318 Mfr # 034055
COMPARE
₹7,400.00INR / Each
Trajan SGE 034056 Replacement Kit, for 1 uL Plunger-in-Needle Syringe, 50 mm L, 0.63 mm OD, BT; 1/Ea 1600319 Mfr # 034056
COMPARE
₹7,400.00INR / Each
Trajan SGE 034057 Replacement Kit, for 1 uL Plunger-in-Needle Syringe, 70 mm L, 0.63 mm OD, CT; 1/Ea 1600320 Mfr # 034057
COMPARE
₹7,400.00INR / Each
Trajan SGE 034060 Replacement Kit, Compatible with 1 uL Plunger-in-Needle Syringe, 70 mm L, 0.63 mm OD, BT; 1/Ea 1600322 Mfr # 034060
COMPARE
₹7,400.00INR / Each
Trajan SGE 035057 Replacement Kit, for 5 uL Plunger-in-Needle Syringe, 70 mm L, 0.63 mm OD, CT; 1/Ea 1600330 Mfr # 035057
COMPARE
₹7,400.00INR / Each
Trajan SGE 035058 Replacement Kit, for 5 uL Plunger-in-Needle Syringe, 70 mm L, 0.63 mm OD, BT; 1/Ea 1600331 Mfr # 035058
COMPARE
₹7,400.00INR / Each
Showing 1- 8 of 8