Trajan SGE 034055 Replacement Kit, Compatible with 1 uL Plunger-in-Needle Syringe, 50 mm L, 0.63 mm OD, CT; 1/Ea
Representative image only.
Trajan SGE 034055 Replacement Kit, Compatible with 1 uL Plunger-in-Needle Syringe, 50 mm L, 0.63 mm OD, CT; 1/Ea
Trajan | Mfr: 034055| Item: 1600318
Inject down to 50nL with high precision and accuracy.
₹7,400.00INR / Each
₹7,400.00INR / Each
Product Description
Key Features
  • Plunger extends into the needle tip
  • Sample contained only in the needle, no glass contact
  • 6 month warranty
More About this Item
Ideal for dispensing very small liquid volumes. When the plunger is depressed, the sample is completely dispensed by the NanoVolume plunger that extends to the tip of the needle.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Kit, Compatible with 1 uL Plunger-in-Needle Syringe, 50 mm L, 0.63 mm OD, CT; 1/Ea