Trajan Gas Chromatography Nuts
Gas Chromatography Nuts for SGE Fittings
  • Configurations to compliment the SGE ferrule product range
  • Usable with tubing sizes ranging from 1/16" to 1/4"
  • Constructed from stainless steel, nickel plated brass, or brass
The design of the nuts and fittings are unique in that the fitting is flat faced and the point of the ferrule is towards the nut. This makes the system more robust since the angled portion of the fitting cannot be damaged.
3,300.00 - ₹5,000.00INR / Pkg of 5
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 103400 Mini Union Nut with 1.2 mm Hole, Stainless Steel; 5/Pk 1599706 Mfr # 103400
COMPARE
₹4,800.00INR / Pkg of 5
Trajan SGE 103405 GC Extended Nut with 1.2 mm Hole, Stainless Steel; 5/Pk 1599710 Mfr # 103405
COMPARE
₹5,000.00INR / Pkg of 5
Trajan SGE 103406 GC Extended Nut with 1.2 mm Hole, Brass(Nickel Plated); 5/Pk 1599711 Mfr # 103406
COMPARE
₹3,300.00INR / Pkg of 5
Trajan SGE 103408 GC Extended Nut with 1/16" Hole, Stainless Steel; 5/Pk 1599713 Mfr # 103408
COMPARE
₹4,800.00INR / Pkg of 5
Trajan SGE 1034060 Brass Nuts, Compatible with Varian Injector; 2/Pk 1599783 Mfr # 1034060
COMPARE
₹3,300.00INR / Pkg of 2