Trajan SGE 103405 GC Extended Nut with 1.2 mm Hole, Stainless Steel; 5/Pk
Representative image only.
Trajan SGE 103405 GC Extended Nut with 1.2 mm Hole, Stainless Steel; 5/Pk
Trajan | Mfr: 103405| Item: 1599710
Gas Chromatography Nuts for SGE Fittings
₹5,000.00INR / Pkg of 5
₹5,000.00INR / Pkg of 5
Product Description
Key Features
  • Configurations to compliment the SGE ferrule product range
  • Usable with tubing sizes ranging from 1/16" to 1/4"
  • Constructed from stainless steel, nickel plated brass, or brass
  • 6 month warranty
More About this Item
The design of the nuts and fittings are unique in that the fitting is flat faced and the point of the ferrule is towards the nut. This makes the system more robust since the angled portion of the fitting cannot be damaged.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE GC Extended Nut with 1.2 mm Hole, Stainless Steel; 5/Pk
  • Warranty6 month warranty