Trajan Fixed-Needle Guided Plunger Syringes
Most rugged syringes available, perfect for industrial environments
  • Extended barrel guides plunger during injection
  • Robust and rugged
Available in 5 µL to 10 µL capacities.
10,700.00 - ₹14,900.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 001450 Microvolume Syringe with Removable Needle and Guided Plunger; 5 uL, 50 mm, 26 G, BT; 1/Ea 1599906 Mfr # 001450
COMPARE
₹12,600.00INR / Each
Trajan SGE 002400 Fixed Needle Guided Plunger Syringe, 10 uL, 50 mm L, 26 Gauge, Bevel Tip; 1/Ea 1599952 Mfr # 002400
COMPARE
₹11,400.00INR / Each
Trajan SGE 002450 Microvolume Syringe with Removable Needle and Guided Plunger; 10 uL, 50 mm, 26 G, BT, 1/Ea 1599953 Mfr # 002450
COMPARE
₹13,800.00INR / Each
Trajan SGE 002455 Gas Tight Guided Plunger with Removable Needle, 10 uL, 50 mm L, 26 gauge; 1/Ea 1599954 Mfr # 002455
COMPARE
₹14,900.00INR / Each
Trajan SGE 001400 Fixed-Needle Guided Plunger Syringe, 5 uL, 50 mm L, 26 Gauge, Bevel Tip; 1/Ea 1601130 Mfr # 001400
COMPARE
₹10,700.00INR / Each