Trajan SGE 001400 Fixed-Needle Guided Plunger Syringe, 5 uL, 50 mm L, 26 Gauge, Bevel Tip; 1/Ea
Representative image only.
Trajan SGE 001400 Fixed-Needle Guided Plunger Syringe, 5 uL, 50 mm L, 26 Gauge, Bevel Tip; 1/Ea
Trajan | Mfr: 001400| Item: 1601130
Most rugged syringes available, perfect for industrial environments
₹10,700.00INR / Each
₹10,700.00INR / Each
Product Description
Key Features
  • Extended barrel guides plunger during injection
  • Robust and rugged
  • 6 month warranty
More About this Item
Available in 5 µL to 10 µL capacities.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Fixed Needle Guided Plunger Syringe, 5 uL, 50 mm L, 26 Gauge, Bevel Tip; 6/Pk