Trajan Fixed Cone-Tip Needle Autosampler Gas Tight Syringes
Designed and tested to meet critical autosampler specifications
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
7,700.00 - ₹49,600.00INR / Each
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
Compareitem Availability Pricing
Trajan SGE 001955 Gas Tight Syringe, Compatible with PerkinElmer Autosampler, 5 uL, 70 mm L, 26 G; 1/Ea 1599923 Mfr # 001955
COMPARE
₹12,300.00INR / Each
Trajan SGE 001957 Gas Tight Syringe, Compatible with PerkinElmer Autosampler, 5 uL, 70 mm L, 23 G; 1/Ea 1599924 Mfr # 001957
COMPARE
₹12,300.00INR / Each
Trajan SGE 002700 Gas Tight Syringe, Compatible with CTC PAL, Fixed CT Needle, 10 uL, 50 mm L, 26 G; 1/Ea 1599956 Mfr # 002700
COMPARE
₹14,000.00INR / Each
Trajan SGE 002812 Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed CT Needle, 10 uL, 23 G; 1/Ea 1599968 Mfr # 002812
COMPARE
₹8,900.00INR / Each
Trajan SGE 002826 Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed CT Needle, 10 uL; 1/Ea 1599976 Mfr # 002826
COMPARE
₹8,800.00INR / Each
Trajan SGE 002827 Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed CT Needle, 10 uL; 6/Pk 1599977 Mfr # 002827
COMPARE
₹49,600.00INR / Pkg of 6
Trajan SGE 002862 Gas Tight Syringe, Compatible with Thermo RSH, Fixed CT Needle, 10 uL, 57 mm L, 23 G; 1/Ea 1599983 Mfr # 002862
COMPARE
₹12,000.00INR / Each
Trajan SGE 002866 Gas Tight Syringe, Compatible with Thermo RSH, Fixed CT Needle, 10 uL, 57 mm L, 26 G; 1/Ea 1599987 Mfr # 002866
COMPARE
₹12,000.00INR / Each
Trajan SGE 002972 Gas Tight Syringe, Compatible with Thermo Scientific TriPlus, Fixed CT Needle, 10 uL; 6/Pk 1600009 Mfr # 002972
COMPARE
₹43,800.00INR / Pkg of 6
Trajan SGE 0029761 Gas Tight Syringe, Compatible with Thermo Scientific Autosamplers, Fixed Cone-Tip Needle,10 uL; 6/Pk 1600011 Mfr # 0029761
COMPARE
₹45,000.00INR / Pkg of 6
Trajan SGE 002977 Gas Tight Syringe, Compatible with CTC/Thermo Autosampler, Fixed Cone-Tip Needle, 10 uL; 1/Ea 1600013 Mfr # 002977
COMPARE
₹7,800.00INR / Each
Trajan SGE 0029771 Gas Tight Syringe, Compatible with Thermo Scientific Autosamplers, 10 uL; 1/Ea 1600014 Mfr # 0029771
COMPARE
₹7,800.00INR / Each
Trajan SGE 002983 Gas Tight Syringe, Compatible with CTC/Thermo Autosampler, Fixed SH Needle, 10 uL; 1/Ea 1600019 Mfr # 002983
COMPARE
₹11,400.00INR / Each
Trajan SGE 002987 Gas Tight Syringe, Compatible with CTC PAL Systems, Fixed Cone-Tip Needle, 10 uL, 23 G; 1/Ea 1600025 Mfr # 002987
COMPARE
₹7,700.00INR / Each
Trajan SGE 0029871 Gas Tight Syringe, Compatible with Thermo Scientific AS2000, Fixed Cone-Tip Needle, 10 uL; 1/Ea 1600026 Mfr # 0029871
COMPARE
₹7,700.00INR / Each
Trajan SGE 003668 Gas Tight Syringe, Compatible with Agilent Autosampler, Fixed Needle, 25 uL, 23-26 G; 1/Ea 1600037 Mfr # 003668
COMPARE
₹14,600.00INR / Each
Trajan SGE 003862 Gas Tight Syringe, Compatible with Thermo RSH, Fixed Needle, 25 uL, 57 mm L, 23 G; 1/Ea 1600043 Mfr # 003862
COMPARE
₹16,700.00INR / Each
Trajan SGE 003866 Gas Tight Syringe, Compatible with Thermo RSH, Fixed Needle, 25 uL, 57 mm L, 26 G; 1/Ea 1600045 Mfr # 003866
COMPARE
₹16,700.00INR / Each
Trajan SGE 003868 Gas Tight Syringe, Compatible with Thermo RSH, Fixed Needle, 25 uL, 85 mm L, 26 G; 1/Ea 1600046 Mfr # 003868
COMPARE
₹20,800.00INR / Each
Trajan SGE 003987 Gas Tight Syringe, Compatible with CTC PAL Systems, Fixed Cone-Tip Needle, 25 uL, 50 mm L, 23 G; 1/Ea 1600047 Mfr # 003987
COMPARE
₹12,800.00INR / Each
Showing 1 - 20 of 34