Trajan SGE 003987 Gas Tight Syringe, Compatible with CTC PAL Systems, Fixed Cone-Tip Needle, 25 uL, 50 mm L, 23 G; 1/Ea
Representative image only.
Trajan SGE 003987 Gas Tight Syringe, Compatible with CTC PAL Systems, Fixed Cone-Tip Needle, 25 uL, 50 mm L, 23 G; 1/Ea
Trajan | Mfr: 003987| Item: 1600047
Designed and tested to meet critical autosampler specifications
₹12,800.00INR / Each
₹12,800.00INR / Each
Product Description
Key Features
  • Plunger design minimizes bending and seizing
  • Cone-tip needle point developed specifically to withstand multiple fast septum injections
  • 6 month warranty
More About this Item
Wide range of fixed and removeable needle options available. PTFE replaceable plunder versions available for use with gas and headspace samples.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • Syringe Capacity (µL)25
  • DescriptionTrajan SGE Gas Tight Syringe, Compatible with CTC PAL, Fixed CT Needle, 25 uL, 50 mm L, 23 G; 1/Ea