Torque Wrenches; Micro Adjustable Click Type
Micro-Adjustable torque wrenches Metal Handle English Scale
The metal handle series offers the user a rugged, industrial strength torque wrench able to withstand the rigors of professional use. Handles are lightly knurled for a tough, non-slip grip. There are 34 models of metal handle micrometer adjustable torque wrenches available.
*Dual scale, calibrated dual direction
*Positive lock with spring loaded pull down lock ring
*Fast, accurate and easy to set
*Easy-to-read laser marked scales
*Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
*Accuracy: +/- 4% clockwise +/- 6% CCW of indicated value, CW, from 20% to 100% of full scale
*Meets or exceeds ASME B107.14M-1994, ISO 6789 Standards
*Individually serialized with matching certificate of calibration traceable to N.I.S.T.
76,500.00 - ₹229,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
J. H. Williams Tool Group 10005MFRMH Micro-Adjustable Click-Type Torque Wrench, 1" Drive, 200-1000 ft lb 1078041 Mfr # 10005MFRMHW
COMPARE
1000 ft lb / 1322 N m
Call For Price
J. H. Williams Tool Group 1002MFRMH Micro-Adjustable Click-Type Torque Wrench, 3/8" Drive, 10-100 ft lb 1078044 Mfr # 1002MFRMHW
COMPARE
100 ft lb / 132.2 N m
₹86,100.00INR / Each
J. H. Williams Tool Group 4004MFRMW Micro-Adjustable Click-Type Torque Wrench, 3/4" Drive, 80-400 ft lb 1078052 Mfr # 4004MFRMHW
COMPARE
400 ft lb / 491 N m
₹229,000.00INR / Each
J. H. Williams Tool Group 1502MRMH Micro-Adjustable Click-Type Torque Wrench, 3/8" Drive, 20-150 in lb 3280201 Mfr # 1502MRMHW
COMPARE
150 in lb / 15.3 N m
₹76,500.00INR / Each
J. H. Williams Tool Group 10002MRMH Micro-Adjustable Click-Type Torque Wrench, 3/8" Drive, 150-1000 in lb 9750345 Mfr # 10002MRMHW
COMPARE
1000 in lb / 110.2 N m
₹78,100.00INR / Each