Tissue Pulverizer
Use for freeze-fracturing tough tissues like skin, cartilage, cornea, hair, nails and fresh bone
  • 316 stainless steel construction designed for liquid nitrogen temperature
This tissue pulverizer reduces samples frozen in liquid nitrogen to a coarse powder. Use this powder as a preliminary step for cell disruption (homogenization) and metabolite, protein and nucleic acid extraction. Available in 2 sizes for tissues samples of 10 mg to 1 g.
117,000.00 - ₹124,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Sample Size (mg) Min Sample Size (mg)
Availability Pricing
BioSpec Products 59012N Tissue Pulverizer, 10 to 100 mg tissue capacity 3690300 Mfr # 59012N
COMPARE
100 10
₹117,000.00INR / Each
BioSpec Products 59013N Tissue Pulverizer, 0.1 to 1 g tissue capacity 3690305 Mfr # 59013N
COMPARE
1000 100
₹124,000.00INR / Each