BioSpec Products 59012N Tissue Pulverizer, 10 to 100 mg tissue capacity
Representative image only.
BioSpec Products 59012N Tissue Pulverizer, 10 to 100 mg tissue capacity
BioSpec Products | Mfr: 59012N| Item: 3690300
Use for freeze-fracturing tough tissues like skin, cartilage, cornea, hair, nails and fresh bone
₹117,000.00INR / Each
₹117,000.00INR / Each
Product Description
Includes
mortar, pestle and polypropylene insulated container.
Key Features
  • 316 stainless steel construction designed for liquid nitrogen temperature
  • 2 year warranty
More About this Item
This tissue pulverizer reduces samples frozen in liquid nitrogen to a coarse powder. Use this powder as a preliminary step for cell disruption (homogenization) and metabolite, protein and nucleic acid extraction. Available in 2 sizes for tissues samples of 10 mg to 1 g.
Two year warranty
Specifications
  • Dispersion TypeCoarse
  • Min Sample Size (g)0.01
  • Min Sample Size (mg)10
  • Max Sample Size (g)0.1
  • Max Sample Size (mg)100
  • MaterialGlass
  • DescriptionTissue Pulverizer, 10 to 100 mg tissue capacity