Tintometer Comparator 2000 Portable Color Grading Systems
Economical, visual color measurement in laboratory, plant, or field
  • Two-Field Comparator system for simple color matching
  • Supplied with lighting unit for standardized measurements
These durable, portable systems allow you to make quick visual comparisons between your sample and a range of stable colored glass standards on a disc. Battery operated illuminator offers flexibility for laboratory, plant, or field color grading. With the two-section field of view, the sample transmittance is directly compared with individual colored glass standards on a test disc.
The comparator is an easy-to-use, entry level color measurement system that compares the color of the sample directly against the color of glass in a disc. Just rotate the disc to get the closest result to the scale. Ideal for light-colored samples with visual color (pathlength up to 40 mm); an illuminator is added to systems for measuring unsaturated samples requiring a longer pathlength.
1,435,000.00 - ₹1,959,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test
Availability Pricing
Lovibond GARDNER Comparator 2000, Gardner Scale 9956240 Mfr # 433340
COMPARE
1 to 18 pH Food and beverage, colored oils
₹1,475,000.00INR / Each
Lovibond AF329 HAZEN Comparator 2000, Hazen Colorscale 9956242 Mfr # 433290
COMPARE
0 to 250 Pt-Co Units Platinum Cobalt Color / APHA Color / Hazen Color
₹1,959,000.00INR / Each
Lovibond 433280 Comparator 2000, Platinum-Cobalt/Hazen/APHA, Low (0 to 70 mg) 9956244 Mfr # 433280
COMPARE
0 to 70 mg Pt/L Water and wastewater, edible oils, petroleum
₹1,482,000.00INR / Each
Lovibond 433250 Comparator 2000, Platinum-Cobalt/Hazen/APHA, High (10 to 250 mg) 9956248 Mfr # 433250
COMPARE
10 to 250 mg Pt/L Water and wastewater, chemicals, edible oils
₹1,435,000.00INR / Each