Lovibond AF329 HAZEN Comparator 2000, Hazen Colorscale
Representative image only.
Lovibond AF329 HAZEN Comparator 2000, Hazen Colorscale
Lovibond | Mfr: 433290| Item: 9956242
Economical, visual color measurement in laboratory, plant, or field
₹1,959,000.00INR / Each
₹1,959,000.00INR / Each
Buy It Together
Current Item
Lovibond AF329 HAZEN Comparator 2000, Hazen Colorscale
Lovibond AF329 HAZEN Comparator 2000, Hazen Colorscale
9956242
₹1,959,000.00INR / Each
+
Lovibond 605960 Replacement Optical Glass Cell, 10 mm
9956251
₹37,800.00INR / Each
=
Total Price 1996800.00 ADD 2 ITEMS TO CART
Product Description
Includes
Nessleriser Daylight 2000+, stand for Nessleriser unit, CAA hazen scale discs (0-30, Pt-Co units), CAB hazen scale discs (30-70, Pt-Co units), NSB hazen scale discs (70-250, Pt-Co units), 2150 Nessleriser body, 2250 Nessleriser body, a pair of Nessler cylinders AF306P, a pair of Nessler cylinders 250mm DB420.
Key Features
  • Two-Field Comparator system for simple color matching
  • Supplied with lighting unit for standardized measurements
  • Color scales include Pt-Co, Hazen, and APHA
  • Pt-Co range: 0 to 250
  • 1 year warranty
More About this Item
These durable, portable systems allow you to make quick visual comparisons between your sample and a range of stable colored glass standards on a disc. Battery operated illuminator offers flexibility for laboratory, plant, or field color grading. With the two-section field of view, the sample transmittance is directly compared with individual colored glass standards on a test disc.
The comparator is an easy-to-use, entry level color measurement system that compares the color of the sample directly against the color of glass in a disc. Just rotate the disc to get the closest result to the scale. Ideal for light-colored samples with visual color (pathlength up to 40 mm); an illuminator is added to systems for measuring unsaturated samples requiring a longer pathlength.
The AF329 is a special kit for visual color grading using Platinum-Cobalt/Hazen/APHA Color over the range 0-250 Pt-Co units. It is typically used for light yellow liquids such as water, oils, and chemicals. Because the sample colors are relatively unsaturated, the kit employs a long path length Nessleriser comparator to give greater measurement sensitivity and accuracy. The Comparator 2000+ is an easy-to-use, entry level color measurement system that compares the color of the sample directly against the color of glass in a disc. All you need to do is rotate the disc to get the closest result to the scale.
1 Year Warranty
Specifications
  • TestPlatinum Cobalt Color / APHA Color / Hazen Color
  • Measurement Points0 to 250 Pt-Co Units
  • DescriptionComparator 2000 for light yellow liquids such as water, oils, and chemicals