ThermoSafe Brands Thick-Walled Shipping Containers
Thick walled for long shipments
These thick-walled shippers are designed for prolonged shipping of temperature-sensitive products. Compared to traditional EPS shippers, these heavy-duty containers allow you to extend shipping times by three days or more when used with dry ice.
Insulated foam container is made from seamless polystyrene with rounded edges. Dome lid is tight-fitting for maximum temperature protection. Containers include corrugated carton.
18,700.00 - ₹22,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Inside Height (in) Inside Length (in) Inside Width (in)
Availability Pricing
ThermoSafe 802 Brands Thick-Walled Shipping Containers, 17-1/4"L x 10"W x 12-3/4"H 0373222 Mfr # 802
COMPARE
12 3/4 17 1/4 10
₹22,100.00INR / Each
ThermoSafe 804 Thick-Walled Shipper, 19.25 x 12.4 x 12.25 in; 1/Pk 0373224 Mfr # 804
COMPARE
12 1/4 19 1/4 12 3/8
₹18,700.00INR / Each