Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces
Electronic controls offer programmable operation
  • General purpose furnace ideal for general laboratory uses—gravimetric analysis, sintering, quantitative analysis and heat treating
Single set point models feature digital temperature control. LED displays chamber temperature continuously; displays set point at touch of a button. Single set point (SSP) models let you monitor both set point and actual chamber temperatures. Choose an 8-segment controller (four ramp and four dwell) with additional programming capabilities, and oven temperature protection relay. Furnaces with 350 cubic-inch chambers include a ceramic shelf to double your capacity. A safety interlock switch disconnects power to heating elements when door is opened. Models 33903-02 and 02053-18 feature a single set point controller with one ramp and one dwell, and an overtemperature protection delay.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Control Type Power (VAC)
Availability Pricing
Thermo Scientific Thermolyne F48010-33 Muffle Furnace, 350 cu in, Single Set Point; 240 VAC, EU/Schuko Plug 0205314 Mfr # F48010-33
COMPARE
5.7 Single set point 240
Call For Price
Thermo Scientific Thermolyne F48020-33 Muffle Furnace, 350 cu in, ramp/dwell control; 240V 0205318 Mfr # F48020-33
COMPARE
5.7 Ramp/dwell 240
Call For Price
Thermo Scientific Thermolyne F48025-60 Muffle Furnace, 350 cu in, ramp/dwell control; 120V 3390302 Mfr # F48025-60
COMPARE
5.7 Ramp/dwell 120