Thermo Scientific Series 8000 Water-Jacketed Incubators
Superior selection, stability, and protection
A. Water-Jacketed Automatic CO2 Incubators
TC sensor for constant temperature and humidity applications
IR sensor for changing temperature and humidity conditions
With a triple-wall construction and large volume of water, these CO2 incubators provide unsurpassed temperature stability and protection against heat loss. The water jacket technology holds the temperature for extended periods of time, which is critical during power failures. They also provide fast temperature recovery.
These incubators feature a temperature-adjustable heated door — keeps inner glass door free of condensation for improved viewing of cultures. Audible and visual alarms indicate temperature deviations of +/- 1°C or more, CO2 concentration deviations or 1% or greater, or when incubator door has been left open for more than five minutes.
Select between thermal conductivity (TC) or infrared (IR ) sensor CO2 incubators. Optional HEPA filter (99.97% efficient at 0.5 microns) can be easily installed to ensure aseptic cell culture; contact Cole-Parmer for more information.
B. Direct Heat CO2 Incubators
Hot air sterilization at 140°C meets GLP/GMP prerequisites
Seamless stainless steel interior minimizes contamination
This incubator is designed to create an optimal simulation of the natural cellular growth environment. Direct heat sterilization cycle — 120 minutes at 140°C — ensures the elimination of all microorganisms and fungal spores from every incubator surface.
Unit features intelligent electronic control of temperature and infrared CO2 measurement resulting in optimal stability and fast recovery times after door openings.
Select between thermal conductivity (TC) or infrared (IR) CO2 incubators. Optional HEPA fitler (99.97% efficient at 0.5 microns) can be easily installed to ensure aseptic cell culture; contact Cole-Parmer for more information.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
Thermo Scientific 3429 WJ Incubator CO2 TC sensor 1595391 Mfr # 3429
COMPARE
Call For Price
Thermo Scientific 3423 WJ Incubator CO2 IR sensor 1595392 Mfr # 3423
COMPARE
Call For Price
Thermo Scientific 3425 WJ Incubator CO2/O2 TC sensor 1595393 Mfr # 3425
COMPARE
Call For Price
Thermo Scientific 3427 WJ Incubator CO2/O2 IR sensor 1595394 Mfr # 3427