Thermo Scientific 3423 WJ Incubator CO2 IR sensor
Representative image only.
Thermo Scientific 3423 WJ Incubator CO2 IR sensor
Thermo Scientific | Mfr: 3423| Item: 1595392
Superior selection, stability, and protection
Call For Price
Call For Price
Product Description
Includes
five stainless steel shelves and gas filter interface assembly.
Key Features
  • 1 year warranty
More About this Item
A. Water-Jacketed Automatic CO2 Incubators
TC sensor for constant temperature and humidity applications
IR sensor for changing temperature and humidity conditions
With a triple-wall construction and large volume of water, these CO2 incubators provide unsurpassed temperature stability and protection against heat loss. The water jacket technology holds the temperature for extended periods of time, which is critical during power failures. They also provide fast temperature recovery.
These incubators feature a temperature-adjustable heated door — keeps inner glass door free of condensation for improved viewing of cultures. Audible and visual alarms indicate temperature deviations of +/- 1°C or more, CO2 concentration deviations or 1% or greater, or when incubator door has been left open for more than five minutes.
Select between thermal conductivity (TC) or infrared (IR ) sensor CO2 incubators. Optional HEPA filter (99.97% efficient at 0.5 microns) can be easily installed to ensure aseptic cell culture; contact Cole-Parmer for more information.
B. Direct Heat CO2 Incubators
Hot air sterilization at 140°C meets GLP/GMP prerequisites
Seamless stainless steel interior minimizes contamination
This incubator is designed to create an optimal simulation of the natural cellular growth environment. Direct heat sterilization cycle — 120 minutes at 140°C — ensures the elimination of all microorganisms and fungal spores from every incubator surface.
Unit features intelligent electronic control of temperature and infrared CO2 measurement resulting in optimal stability and fast recovery times after door openings.
Select between thermal conductivity (TC) or infrared (IR) CO2 incubators. Optional HEPA fitler (99.97% efficient at 0.5 microns) can be easily installed to ensure aseptic cell culture; contact Cole-Parmer for more information.
1 Year Warranty
Specifications
  • DescriptionWJ Incubator CO2 IR sensor