Thermo Scientific Orion Star A200 Series Conductivity/TDS/Salinity/Resistivity/Temperature Meter
Durability and dependability make it the ideal choice for every sample, every day, anywhere
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Meets all US Pharmacopoeia (USP) requirements
  • Selectable cell constant for use with 2- or 4- cell conductivity probes
  • Improved stability algorithm provides better measurement accuracy
  • Selectable reference temperature of 5, 10, 15, 20, or 25°C
  • Nonvolatile memory holds 1000 data points with time date stamp
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A222 conductivity, TDS, salinity, resistivity and temperature meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD shows measurement and temperature along with electrode ststus, time/date, sample ID, user ID, and calibration data.
GLP-related features include 1000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only or kit configurations.
157,380.00 - ₹247,740.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A222 Star A222 Conductivity Portable Meter Only 1970416 Mfr # STARA2220
COMPARE
Star A222 Conductivity Portable Meter Only
₹157,380.00INR / Each
Thermo Scientific STAR A222 Star A222 Conductivity Portable Meter Kit 1970418 Mfr # STARA2225
COMPARE
Star A222 Conductivity Portable Meter Kit
₹247,740.00INR / Each