Thermo Scientific Orion Star A200 DO/RDO/Temperature Meter
Durability and dependability make it the ideal choice for every sample, every day, anywhere
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Choose between RDO optical probe and polarographic probe
  • Directly measure in mg/L (ppm) or % saturation
  • Automatic temperature, barometric pressure abd salinity corrections
  • Nonvolatile emeory holds up to 1000 data points with time date stamp
  • Four AA batteries provide 800 hours of field use, or order optional AC adpater for lab use
Thermo Scientific Orion Star A223 dissolved oxygen meter offers the option of RDO optical probes or traditional polarographic probes. Meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD shows measurement with temperature, along with electrode status, time/date stamp, sample ID, user ID, and calibration data.
GLP related features include 1000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only or kit configurations.
154,920.00 - ₹316,740.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STARA2230 Star A223 DO Portable Meter Only 5310116 Mfr # STARA2230
COMPARE
Star A223 DO Portable Meter Only
₹154,920.00INR / Each
Thermo Scientific STAR A223 Star A223 DO Portable Meter Kit 5310118 Mfr # STARA2235
COMPARE
Star A223 DO Portable Meter Kit
₹316,740.00INR / Each