Thermo Scientific STARA2230 Star A223 DO Portable Meter Only
Representative image only.
Thermo Scientific STARA2230 Star A223 DO Portable Meter Only
Thermo Scientific | Mfr: STARA2230| Item: 5310116
Durability and dependability make it the ideal choice for every sample, every day, anywhere
₹154,920.00INR / Each
₹154,920.00INR / Each
Buy It Together
Current Item
Thermo Scientific STARA2230 Star A223 DO Portable Meter Only
Thermo Scientific STARA2230 Star A223 DO Portable Meter Only
5310116
₹154,920.00INR / Each
+ =
Total Price 307480.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Choose between RDO optical probe and polarographic probe
  • Directly measure in mg/L (ppm) or % saturation
  • Automatic temperature, barometric pressure abd salinity corrections
  • Nonvolatile emeory holds up to 1000 data points with time date stamp
  • Four AA batteries provide 800 hours of field use, or order optional AC adpater for lab use
  • 3 year warranty
More About this Item
Thermo Scientific Orion Star A223 dissolved oxygen meter offers the option of RDO optical probes or traditional polarographic probes. Meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD shows measurement with temperature, along with electrode status, time/date stamp, sample ID, user ID, and calibration data.
GLP related features include 1000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only or kit configurations.
Meter only includes: four AA batteries.
3 Year WarrantyIP 67ISO9001:2000 Certified Supplier
Specifications
  • Meter TypePortable
  • Min Dissolved Oxygen0 to 90 ppm; 0 to 500% saturation
  • Max Temperature (° C)105
  • Probe IncludedNo
  • DescriptionStar A223 DO Portable Meter Only