Thermo Scientific Orion Star A100 Series Portable Conductivity/TDS/Temperature Meter
Simple, accurate conductivity measurements
  • Match range with selectable cell constant from 0.1 to 10.0K
  • Increase accuracy with selectable reading reference temperature of 20°C or 25°C
  • One-point manual or automatic calibration
  • Large LCD displays Conductivity along with temperature
  • Auto-Read™ function locks in stable reading
  • Nonvolatile memory holds up to 50 data points
  • IP67-rated housing
Thermo Scientific Orion Star A122 portable conductivity meter combines simplicity with accuracy. Large informative display shows conductivity or TDSreadings, temperature, meter mode, calibration information, and battery life. Simple button layout and on-screen messages help with calibration and setup menu choices. Waterproof and protected to take anywhere with an IP67-rated housing.
Auto-Read™ function locks in stable reading and nonvolatile memory holds up to 50 data points. Four AA batteries (included) provide over 2000 hours of operation. Universal power adapter sold separately to run meter on AC power.
122,040.00 - ₹222,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A122 Star A122 Conductivity Portable Meter 1970445 Mfr # STARA1220
COMPARE
Star A122 Conductivity Portable Meter
₹122,040.00INR / Each
Thermo Scientific STAR A122 Star A122 Conductivity Portable Meter Kit 1970447 Mfr # STARA1225
COMPARE
Star A122 Conductivity Portable Meter Kit
₹222,100.00INR / Each