Thermo Scientific Orion Green pH Electrodes
Environmentally-friendly line of pH electrodes help make the you and the world a little safer!
  • Made without mercury or lead - safe to use and easy to dispose!
  • Great replacement for calomel electrodes that contain mercury
Orion green electrodes have a durable epoxy body that can be used under rugged conditions. Use to read pH from 0 to 14. Choose low-maintenance gel-filled or high-accuracy, reusable refillable electrodes, depending on your application and budget. Available with single-junction reference for routine pH measurement or double-junction for use with TRIS buffers, proteins, and sulfides.
Waterproof BNC connectors provide a waterproof and secure connection when used with an Orion Star™ Series meter. This BNC connector is compatible with any pH meter with standard BNC input.
33,930.00 - ₹47,770.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Electrode Body Material Reference Junction
Availability Pricing
Thermo Scientific GS9106BNWP Green pH Probe, Sealed/SJ/Epoxy; WP BNC 0571400 Mfr # GS9106BNWP
COMPARE
Sealed Epoxy Glass capillary
₹33,930.00INR / Each
Thermo Scientific GD9156BNWP Green pH Probe, Refillable/DJ/Epoxy; WP BNC 0571406 Mfr # GD9156BNWP
COMPARE
Refillable Epoxy Glass fiber
₹47,770.00INR / Each