Thermo Scientific GS9106BNWP Green pH Probe, Sealed/SJ/Epoxy; WP BNC
Representative image only.
Thermo Scientific GS9106BNWP Green pH Probe, Sealed/SJ/Epoxy; WP BNC
Thermo Scientific | Mfr: GS9106BNWP| Item: 0571400
Environmentally-friendly line of pH electrodes help make the you and the world a little safer!
₹33,930.00INR / Each
₹33,930.00INR / Each
Product Description
Includes
3.3-ft (1-m) cable with BNC connector. Refillable electrodes include 60-mL bottle of electrode filling solution.
Key Features
  • Made without mercury or lead - safe to use and easy to dispose!
  • Great replacement for calomel electrodes that contain mercury
  • 6 month warranty
More About this Item
Orion green electrodes have a durable epoxy body that can be used under rugged conditions. Use to read pH from 0 to 14. Choose low-maintenance gel-filled or high-accuracy, reusable refillable electrodes, depending on your application and budget. Available with single-junction reference for routine pH measurement or double-junction for use with TRIS buffers, proteins, and sulfides.
Waterproof BNC connectors provide a waterproof and secure connection when used with an Orion Star™ Series meter. This BNC connector is compatible with any pH meter with standard BNC input.
6 Month Warranty
Specifications
  • ApplicationsSealed
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGlass capillary
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)90
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • Cable Length (m)1
  • DescriptionGreen pH Probe, Sealed/SJ/Epoxy; WP BNC
  • Warranty6 month warranty