Thermo Scientific™ Orion™ Automatic Temperature Compensation Probes
Provides ATC with for Orion pH meters
  • Locking connector prevents lost data and probes
  • Choose the right material for your application
  • Optional adapter cables available for older Orion models
Available in a variety of materials and sizes to provide reliable temperature compensated pH measurement across a wide range of applications. Proprietary waterproof MiniDIN locking connector prevents lost data or even lost probes when used with Orion Star Series meters. Probes include a 3.3-ft (1-m) cable.
Order optional adapter cables to connect probes to older Orion meters separately.
55,720.00 - ₹144,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific 927007MD Stainless Steel Automatic Temperature Compensation Probe 5882448 Mfr # 927007MD
COMPARE
Stainless Steel Automatic Temperature Compensation Probe
₹55,720.00INR / Each
Thermo Scientific 928007MD Micro Epoxy/SS Automatic Temperature Compensation Probe 5882450 Mfr # 928007MD
COMPARE
Micro Epoxy/SS Automatic Temperature Compensation Probe
₹144,000.00INR / Each
Thermo Scientific 927005MD Epoxy Automatic Temperature Compensation Probe 5882454 Mfr # 927005MD
COMPARE
Epoxy Automatic Temperature Compensation Probe
₹114,000.00INR / Each
Thermo Scientific 927006MD Glass Automatic Temperature Compensation Probe 5882458 Mfr # 927006MD
COMPARE
Glass Automatic Temperature Compensation Probe
₹118,000.00INR / Each