Thermo Scientific 927007MD Stainless Steel Automatic Temperature Compensation Probe
Representative image only.
Thermo Scientific 927007MD Stainless Steel Automatic Temperature Compensation Probe
Thermo Scientific | Mfr: 927007MD| Item: 5882448
Provides ATC with for Orion pH meters
₹55,720.00INR / Each
₹55,720.00INR / Each
Product Description
Key Features
  • Locking connector prevents lost data and probes
  • Choose the right material for your application
  • Optional adapter cables available for older Orion models
  • 3 year warranty
More About this Item
Available in a variety of materials and sizes to provide reliable temperature compensated pH measurement across a wide range of applications. Proprietary waterproof MiniDIN locking connector prevents lost data or even lost probes when used with Orion Star Series meters. Probes include a 3.3-ft (1-m) cable.
Order optional adapter cables to connect probes to older Orion meters separately.
3 Year Warranty
Specifications
  • Length (mm)120
  • Diameter (mm)6
  • DescriptionStainless Steel Automatic Temperature Compensation Probe