Thermo Scientific™ Nunc™ Thermanox™ Coverslips
Cell culture treated on one side for optimal cell growth and attachment
  • Flat and easy-to-handle—0.2 mm thickness
  • Thermanox® plastic is resistant to solvents commonly used in electron microscopy
  • Thermanox® exhibits autoflourescence in the range of 380 to 545 nm
Improve cell attachment and growth on coverslips with the versatile Thermo Scientific™ Nunc™ Thermanox™ Coverslips. These cell-culture treated coverslips are available in a number of shapes and sizes. The proprietary polyester material is highly resistant to solvents such as xylene, acetone, and acetic acid, and has a very low gas permeability. Suitable for cytostaining, mounting, embedding, or scintillation counting. Not recommended for phase contrast microscopy or techniques involving fluorescent stains.
These coverslips are safe, easy to handle, and radiation sterilized.
171,000.00 - ₹253,000.00INR / Case of 500
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Size Thickness Description
Availability Pricing
Thermo Scientific Nunc 174950 Coverslips, Plastic, Sterile, Round, 13 mm Diameter; 500/Cs 0183900 Mfr # 174950
COMPARE
13 mm Diameter 0.2 mm Coverslips, Plastic, Sterile, Round, 13 mm Diameter; 500/Cs
₹175,000.00INR / Case of 500
Thermo Scientific Nunc 174985 Coverslips, Plastic, Sterile, Round, 25 mm Diameter; 500/Cs 0183901 Mfr # 174985
COMPARE
25 mm Diameter 0.2 mm Coverslips, Plastic, Sterile, Round, 25 mm Diameter; 500/Cs
₹177,000.00INR / Case of 500
Thermo Scientific Nunc 174942 Coverslips, Plastic, Sterile, Rectangular, 60 mm L x 22 mm W; 500/Cs 0183902 Mfr # 174942
COMPARE
60 mm L x 22 mm W 0.2 mm Coverslips, Plastic, Sterile, Rectangular, 60 mm L x 22 mm W; 500/Cs
₹253,000.00INR / Case of 500
Thermo Scientific Nunc 174969 Coverslips, Plastic, Sterile, Round, 15 mm Diameter; 500/Cs 0183905 Mfr # 174969
COMPARE
15 mm Diameter 0.2 mm Coverslips, Plastic, Sterile, Round, 15 mm Diameter; 500/Cs
₹171,000.00INR / Case of 500
Thermo Scientific Nunc 174977 Coverslips, Plastic, Sterile, Round, 22 mm Diameter; 500/Cs 0183906 Mfr # 174977
COMPARE
22 mm Diameter 0.2 mm Coverslips, Plastic, Sterile, Round, 22 mm Diameter; 500/Cs
₹177,000.00INR / Case of 500