Thermo Scientific NESLAB ThermoFlex Recirculating Chillers

Up to 10% greater cooling capacity than comparable models
  • High reliability
  • Easy to operate and install
These reliable, easy-to-use chillers are optimized for the most demanding applications. They feature a robust refrigeration system designed for continuous use in a variety of environments. The recirculation system incorporates a highly reliable plumbing design to eliminate leaks and improve performance. Integrated air and fluid filters minimize wear to mechanical parts, which lowers lifetime maintenance costs and improves system reliability.

Units feature simple push-button operation with a quick-start guide to walk you through start-up. A visual fluid indicator gives instant status of fluid level while an integrated funnel allows quick, spill-proof filling. Integrated ramp and locking casters allow a single person to unpack, move, and install the chiller in minutes. Unit can be operated with two sides blocked for installation flexibility. Chillers are available with low-flow (P1) or high-flow (T1) positive displacement pump with Internal Pressure Regulator (IPR) to protect your application from excessively high pressures.
2,031,000.00INR / Each

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 20° C (watts) Min Temperature (° C) Description
Availability Pricing
Thermo Scientific 1011C10500200100 ThermoFlex 900 Chiller, P1; 115V WW-13042-00 Mfr # 1011C10500200100
COMPARE
900 5 900 Chiller, P1; 115V
₹2,031,000.00INR / Each
Thermo Scientific 1011C10500200100 ThermoFlex 900 Chiller, P1, IPR; 115 VAC WW-13042-04 Mfr # 1011C10500200100
COMPARE
900 5 900 Chiller, P1, IPR; 115V
Call For Price
Thermo Scientific 1011C10500200100 ThermoFlex 900 Chiller, T1, IPR; 115 VAC WW-13042-10 Mfr # 1011C10500200100
COMPARE
900 5 900 Chiller, P2, IPR; 115V
Call For Price
Thermo Scientific 1014C10520200500 ThermoFlex 900 Chiller, P1, IPR; 230 VAC WW-13042-16 Mfr # 1014C10520200500
COMPARE
750 5 900 Chiller, P1, IPR; 230V
Call For Price
Thermo Scientific 1111C10500200100 ThermoFlex 1400 Chiller, P1, IPR; 115 VAC WW-13042-34 Mfr # 1111C10500200100
COMPARE
1400 5 1400 Chiller, P1, IPR; 115V
Call For Price
Thermo Scientific 1114C10500210400 ThermoFlex 1400 Chiller, P1, IPR; 230 VAC WW-13042-36 Mfr # 1114C10500210400
COMPARE
1170 5 1400 Chiller, P1, IPR; 230V
Call For Price
Thermo Scientific 1213C10500200200 ThermoFlex 2500 Chiller, P1, IPR; 208 VAC WW-13042-42 Mfr # 1213C10500200200
COMPARE
2500 5 2500 Chiller, P1, IPR; 208V
Call For Price
Thermo Scientific 1213C10500200200 ThermoFlex 2500 Chiller, T1, IPR; 208 VAC WW-13042-46 Mfr # 1213C10500200200
COMPARE
2500 5 2500 Chiller, P2, IPR; 208V
Call For Price
Thermo Scientific 1313110520200200 ThermoFlex 3500 Chiller, T1, IPR; 208 VAC WW-13042-54 Mfr # 1313110520200200
COMPARE
3500 5 3500 Chiller, P2, IPR; 208V
Call For Price
Thermo Scientific 1413110500200B00 ThermoFlex 5000 Chiller, T1, IPR; 208 VAC WW-13042-58 Mfr # 1413110500200B00
COMPARE
5000 5 5000 Chiller, P2, IPR; 208V
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat