Thermo Scientific Nalgene Round Fluorinated Carboys with Grips, FLPE
Minimize solvent absorption and pentration
  • Won't leak or puncture
  • Graduated in 1 gal and 5L increments
Fluorination provides economical and effective barrier properties and reduces solvent absorption and penetration on all surfaces. Leakproof and puncture resistant, these narrow-mouthed carboys are graduated in 1-gallon and 5-liter increments.
37,800.00 - ₹52,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2097-0020 fluorinated carboy with grip, 10 L 6250050 Mfr # 2097-0020
COMPARE
10 83B Fluorinated High Density Polyethylene (FLPE)
₹37,800.00INR / Each
Thermo Scientific Nalgene 2097-0050 Round Fluorinated Carboys with Grips, 20 L 6250060 Mfr # 2097-0050
COMPARE
20 83B Fluorinated High Density Polyethylene (FLPE)
₹52,100.00INR / Each