Thermo Scientific Nalgene Polycarbonate Carboys with Spigot
Ideal for refrigerated or frozen storage of aqueous solutions
  • More durable, lighter & safer than glass
  • Clear for visual inspection of contents
  • Autoclavable for sterile applications
Polycarbonate (PC) carboys are graduated in 1-gallon and 5-liter increments. Polypropylene (PP) cap with TPE gasket ensures leakproof seal. The PP spigot accepts 5/8" ID tubing and comes with adapter for 1/4" and 1/2" ID tubing. Autoclavable.
59,900.00 - ₹77,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Capacity (L) Cap / Stopper Size
Availability Pricing
Thermo Scientific Nalgene 2317-0020 Polycarbonate Carboy with Spigot; 10 L WW-06063-80 Mfr # 2317-0020
COMPARE
Yes 10 83B
₹59,900.00INR / Each
Thermo Scientific Nalgene 2317-0050 Polycarbonate Carboy with Spigot; 20 L WW-06063-82 Mfr # 2317-0050
COMPARE
Yes 20 83B
₹77,300.00INR / Each