Thermo Scientific Nalgene Polycarbonate Carboy
Ideal for refrigerated or frozen storage of aqueous solutions
  • More durable, lighter & safer than glass
  • Clear for visual inspection of contents
  • Autoclavable for sterile applications
Polycarbonate (PC) carboys are graduated in 1-gallon and 5-liter increments. Polypropylene (PP) cap with TPE gasket ensures leakproof seal. Autoclavable.
63,300.00 - ₹67,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2251-0020 polycarbonate carboy, 10 L WW-06063-60 Mfr # 2251-0020
COMPARE
10 83B Polycarbonate(PC)
₹67,000.00INR / Each
Thermo Scientific Nalgene 2251-0050 polycarbonate carboy, 20 L WW-06063-62 Mfr # 2251-0050
COMPARE
20 83B Polycarbonate(PC)
₹63,300.00INR / Each