Thermo Scientific™ Nalgene™ PMP Beakers with Handle
Sturdy easy-grip handles
  • Molded in graduation in both oz. and mL won't wear off with continuous use or washings
  • Built-in rectangular handle provides easy, safe handling and pouring
  • Autoclavable
Ensure resistance against acids, bases, aliphatic alcohols, aldehydes and esters with Thermo Scientific™ Nalgene™ polymethylpentene (PMP) graduated beakers with handles. These beakers have rectangular handles for easy carrying and pouring and can withstand continuous use at high temperatures.
These beakers withstand continuous exposure to temperatures as high as 150°C and intermittent temperatures to 175°C (347°F).
20,300.00 - ₹23,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 1223-1000 PMP Graduated Beaker w/ Handle, 1000 mL 0600497 Mfr # 1223-1000
COMPARE
1000 200 Polymethylpentene
₹20,300.00INR / Each
Thermo Scientific Nalgene 1223-3000 PMP Graduated Beaker w/ Handle, 3000 mL 0600499 Mfr # 1223-3000
COMPARE
3000 400 Polymethylpentene
₹23,500.00INR / Each