Thermo Scientific™ Nalgene™ HDPE Beakers with Handle
Sturdy easy-grip handles
  • Molded in graduation in both oz. and mL won't wear off with continuous use or washings
  • Built-in rectangular handle provides easy, safe handling and pouring
Safely handle acids, alkalis and hot solutions using Thermo Scientific™ Nalgene™ Graduated HDPE Beakers with Handles. These high density polyethylene beakers feature an extra-heavy wall and a sturdy rectangular handle for reliable support and dispensing control.
17,600.00 - ₹18,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Thermo Scientific Nalgene 1220-1250 HDPE Graduated Beaker w/ Handle, 1000 mL 0600510 Mfr # 1220-1250
COMPARE
1000 100 High Density Polyethyene (HDPE)
₹17,600.00INR / Each
Thermo Scientific Nalgene 1220-2500 HDPE Graduated Beaker w/ Handle, 3000 mL 0600520 Mfr # 1220-2500
COMPARE
3000 200 High Density Polyethyene (HDPE)
₹18,200.00INR / Each