Thermo Scientific Nalgene Fluorinated Rectangular Carboys with Spigot
Don't waste valuable bench space
  • Space-saving design for no dead area like traditional round carboys
  • Leakproof HDPE for safe transport
  • Liter/gallon graduations for easy volume determination
Surface fluorination on these polyethylene (PE) carboys reduces solvent absorption and permeation. Both feature molded graduations in liters and gallons and a stainless steel handle. Model 62502-10 has sturdy hand grips. Replacement spigot, a 100-mm fluorinated polypropylene closure that accepts 5⁄8" ID tubing, is made of Tefzel® ETFE. UN-approved.
65,900.00 - ₹69,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene DS2327-0020 fluorinated carboy with spigot, 9 L 6250200 Mfr # DS2327-0020
COMPARE
9 70 mm Polyethylene(PE)
₹65,900.00INR / Each
Thermo Scientific Nalgene DS2327-0050 fluorinated carboy with spigot, 20 L 6250210 Mfr # DS2327-0050
COMPARE
20 70 mm Polyethylene(PE)
₹69,400.00INR / Each