Thermo Scientific Nalgene CryoCane Cryogenic Storage Canes
Offer convenient storage and retrieval of vials in cryogenic storage dewars
These aluminum storage canes accommodate a variety of vial sizes and designs. Stepped cane 03755-06 holds five cryogenic vials (1.0, 1.2, 1.5, and 2.0 mL). Standard cane 03755-07 holds six cryogenic vials (1.0, 1.2, 1.5, 2.0, and 5.0 mL).
49,200.00INR / Case of 48
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held
Availability Pricing
Thermo Scientific Nalgene 5015-0001 Cryocanes, Stepped Al, 5 Vials; 48/Cs 0375506 Mfr # 5015-0001
COMPARE
5
₹49,200.00INR / Case of 48
Thermo Scientific Nalgene 5015-0002 Cryocanes, Standard Al, 6 Vials; 48/Cs 0375507 Mfr # 5015-0002
COMPARE
6
₹49,200.00INR / Case of 48