Thermo Scientific Nalgene 5015-0001 Cryocanes, Stepped Al, 5 Vials; 48/Cs
Representative image only.
Thermo Scientific Nalgene 5015-0001 Cryocanes, Stepped Al, 5 Vials; 48/Cs
Thermo Scientific | Mfr: 5015-0001| Item: 0375506
Offer convenient storage and retrieval of vials in cryogenic storage dewars
₹49,200.00INR / Case of 48
₹49,200.00INR / Case of 48
Buy It Together
Current Item
Thermo Scientific Nalgene 5015-0001 Cryocanes, Stepped Al, 5 Vials; 48/Cs
Thermo Scientific Nalgene 5015-0001 Cryocanes, Stepped Al, 5 Vials; 48/Cs
0375506
₹49,200.00INR / Case of 48
+ =
Total Price 76600.00 ADD 2 ITEMS TO CART
Product Description
More About this Item
These aluminum storage canes accommodate a variety of vial sizes and designs. Stepped cane 03755-06 holds five cryogenic vials (1.0, 1.2, 1.5, and 2.0 mL). Standard cane 03755-07 holds six cryogenic vials (1.0, 1.2, 1.5, 2.0, and 5.0 mL).
Specifications
  • Tubes Held5
  • Tube Size1.0, 1.2, 1.5, and 2.0 mL
  • DescriptionCryocanes, Stepped Al, 5 Vials; 48/Cs