Thermo Scientific Nalgene Aspirator Pump
Integral check valve maintains up to 28.5" Hg even when water supply is interrupted
  • Simple installation attaches to standard faucet connection with 3/8" NPT
  • Conserves water with 7.5 psig water pressure
  • Free air capacity is 11.5 LPM when the water supply flow rate is 6.5 LPM
  • Polypropylene construction with 4-3/4" polyethylene discharge tubing
Perfect for basic laboratory vacuum needs, such as aspiration and filtration. System tubing connection is a detachable stepped hose barb that accepts 1/4" to 3/8" inner diameter flexible tubing. Integral check valve will also prevent backflow of water into the vacuum line.
7,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Max Vacuum (torr)
Availability Pricing
Thermo Scientific 6140-0010 Faucet Aspirator/Vacuum Pump; 28.5"Hg 0637830 Mfr # 6140-0010
COMPARE
0.4 28.5 36.1
₹7,100.00INR / Each