Thermo Scientific Matrix Electronic Pipettors
Lightweight and comfortable with intuitive programming
  • Ergonomic design
  • Trigger based operation and tip ejection
  • Step based programming
  • On board memory saves six programs (up to 40 steps eac
  • Advanced battery technology has no memory effect
  • No downtime during recharging, pipettor can be used while plugged in
  • Stepper motor technology and error correction software
The intuitive, easy-to-learn, and easy-to-use step-based programming allows you more time for pipetting and less time on programming. All functions can be sequentially linked together (up to 40 steps) and saved to increase your efficiency and repeatability in your pipetting routines.
Single channel electronic pipettors
Five volume ranges available to meet all your pipetting needs. Additionally, the Shorty version is lighter and it's short barrel makes working in a hood easier. Easily create programs using the logical, intuitive step-based programming which allows the electronic pipettor to perform all tasks you can do with a manual pipettor — only faster and easier.For example, you could aspirate 50 µL and then dispense 10 µL, 10 µL, 10 µL, and 20 µL utilizing specific dispense speeds for each.
Multichannel electronic pipettors
Select from 8-, 12-, or 16-channel models each with large volume capacity — up to 1250 µL per tip. Easily create programs using the logical, intuitive step-based programming which allows the electronic pipettor to perform all tasks you can do with a manual pipettor — only faster and easier. For example, you could aspirate 1200 µL and then dispense 50 µL to the first two rows of a microplate, then 100 µL to the next ten rows utilizing specific dispense speeds for each.
EXP multichannel electronic pipettors
This unique electronic multichannel pipettor allows you to increase productivity with its innovative exandable tip spacing system. Simply pull the rod to expand the tips to access test tube racks, 24-well microplates, or 48-well microplates, for example, and then push the rod to compress the spacing to pipette into 96-well microplates. The EXP Pipettors share the same programming functionality and large tip capacity as the Multichannel Pipettors.
Equalizer multichannel electronic pipettors
This unique electronic multichannel pipettor allows you to increase productivity with its innovative exandable, equal, and stepped tip spacing system. The equal tip spacing expands or contracts with the adjustment of the slide rod to perform sample transfers between virtually any tube rack, microplate or horizontal gel box. The pipettor allows plate-to-plate transfers between 96- and 384-well microplates.
The Equalizer Pipettors share the same programming functionality and large tip capacity as the EXP and Multichannel Pipettors.
83,600.00 - ₹278,000.00INR / Each
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
Thermo Scientific Matrix 1029 Electronic Single Channel Pipettor, 0.5 to 12.5 µL 9499900 Mfr # 1029
COMPARE
0.0125 0.0005
₹83,600.00INR / Each
Thermo Scientific Matrix 1020 Electronic Single Channel Pipettor, 1.0 to 30 µL 9499901 Mfr # 1020
COMPARE
0.03 0.001
₹83,600.00INR / Each
Thermo Scientific Matrix 72405-102130 Phenyl Column - Length 100mm, 2.1mm ID, 5µm particle size 9499902 Mfr # 1021
COMPARE
0.125 0.002
₹83,600.00INR / Each
Thermo Scientific Matrix 1022 Electronic Single Channel Pipettor, 5 to 250 µL 9499903 Mfr # 1022
COMPARE
0.25 0.005
₹83,600.00INR / Each
Thermo Scientific Matrix 1024 Electronic Single Channel Pipettor, 15 to 1250 µL 9499904 Mfr # 1024
COMPARE
1.25 0.015
₹83,600.00INR / Each
Thermo Scientific Matrix 112250 Sterile Nonvented HDPE Caps; 250/Pk, 2Pk/Cs 9499905 Mfr # 1122
COMPARE
0.25 0.005
₹83,600.00INR / Each
Thermo Scientific Matrix 1124 Electronic Single Channel Shorty Pipettor, 15 to 1250 µL 9499906 Mfr # 1124
COMPARE
1.25 0.015
₹83,600.00INR / Each
Thermo Scientific Matrix 2009 Electronic Multichannel Pipettor, 0.5 to 12.5 µL, 8 Channel 9499910 Mfr # 2009
COMPARE
0.0125 0.0005
₹189,000.00INR / Each
Thermo Scientific Matrix 46200101 Electronic Pipette, 1-10 µL, 120V 9499911 Mfr # 2001
COMPARE
0.125 0.002
₹189,000.00INR / Each
Thermo Scientific Matrix 2002-0004 high-density polyethylene narrow-mouth bottle, 4 oz 9499912 Mfr # 2002
COMPARE
0.25 0.005
₹189,000.00INR / Each
Thermo Scientific Matrix 2004-9025 Amber Narrow-mouth HDPE Bottles, 8 mL, 12/Pk 9499913 Mfr # 2004
COMPARE
1.25 0.015
₹189,000.00INR / Each
Thermo Scientific Matrix 2019-1000 Sterile PETG Media Bottles, 1000 mL; 24/box 9499914 Mfr # 2019
COMPARE
0.0125 0.0005
₹213,000.00INR / Each
Thermo Scientific Matrix 2011 Electronic Multichannel Pipettor, 2 to 125 µL, 12 Channel 9499915 Mfr # 2011
COMPARE
0.125 0.002
₹213,000.00INR / Each
Thermo Scientific Matrix 020125 110 VAC adapter for conductivity/salinity meters 9499916 Mfr # 2012
COMPARE
0.25 0.005
₹213,000.00INR / Each
Thermo Scientific Matrix 2014 Electronic Multichannel Pipettor, 15 to 1250 µL, 12 Channel 9499917 Mfr # 2014
COMPARE
1.25 0.015
₹213,000.00INR / Each
Thermo Scientific Matrix 2069 Electronic Multichannel Pipettor, 0.5 to 12.5 µL, 16 Channel 9499918 Mfr # 2069
COMPARE
0.0125 0.0005
₹256,000.00INR / Each
Thermo Scientific Matrix 228103220602 Immersion Cooler -60 to -20C with R probe - 115V 9499919 Mfr # 2060
COMPARE
0.03 0.001
₹256,000.00INR / Each
Thermo Scientific Matrixexp 2061 Electronic Multichannel Pipettor, 2 to 125 µL, 16 Channel 9499920 Mfr # 2061
COMPARE
0.125 0.002
₹256,000.00INR / Each
Thermo Scientific Matrixexp 2621 Electronic Multichannel Pipettor, 2to125µL,6Channel 9499930 Mfr # 2621
COMPARE
0.125 0.002
₹250,000.00INR / Each
Thermo Scientific Matrixexp 262212032000 Recirculating Chiller 13401-50 9499931 Mfr # 2622
COMPARE
0.25 0.005
₹250,000.00INR / Each
Showing 1 - 20 of 34