Thermo Scientific Lindberg Blue M Muffle Furnaces
Rise times as fast as 15 minutes!
  • Microprocessor-based PID control with self-tuning capability to prevent overshooting of set temperature.
Choose from models with either a single set point or a sixteen-segment programmable controller. Safety features include adjustable, high-limit overtemperature protection and a safety interlock system to cut off power to the heating elements when the door is opened.
Energy-efficient MOLDATHERM® insulation and double-wall construction help maintain a cool outer temperature as well as uniform heat distribution within the chamber. An air vent ventilates chamber; an atmosphere inlet lets you purge inert gases.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Control Type Power (VAC)
Availability Pricing
Thermo Scientific Lindberg BF51748C-1 Blue M Muffle Furnace, 121 cu in; 208-240V 3385214 Mfr # BF51748C-1
COMPARE
2 Single set point 208 / 240
Call For Price
Thermo Scientific Lindberg BF51766C-1 Blue M Muffle Furnace, 324 cu in; 208-240V 3385222 Mfr # BF51766C-1
COMPARE
5.3 Single set point 208 / 240
Call For Price
Thermo Scientific Lindberg BF51794C-1 Blue M Box Furnace, 1123 cu in, SSP; 208V 3385227 Mfr # BF51794C-1
COMPARE
18.4 Single set point 208 / 240
Call For Price
Thermo Scientific Lindberg BF51894C-1 Blue M Box Furnace, 1123 cu in, 16-Pt Prog; 208V 3385245 Mfr # BF51894C-1
COMPARE
18.4 16-segment programmable 208 / 240
Call For Price
Thermo Scientific Lindberg BF51828C-1 Blue M Muffle Furnace, 2592 cu in; 208-240V 3385255 Mfr # BF51828C-1
COMPARE
42.5 16-segment programmable 208 / 240